Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015258.1 | NP_056073.1 | 6 | 600 | ||
XM_006717018.2 | XP_006717081.1 | 6 | 517 | |||
XM_006717019.1 | XP_006717082.1 | 5 | 448 | |||
XM_017014552.1 | XP_016870041.1 | 5 | 448 | |||
GenBank mRNA | AB014574.1 | - | 6 | 527 | ||
AK303074.1 | - | 5 | 448 | |||
AK307498.1 | - | 5 | 429 | |||
AK307507.1 | - | 10 | 1034 | |||
BC009609.1 | - | 6 | 557 |