Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015258.1 | NP_056073.1 | 28 | 4199 | ||
XM_006717018.2 | XP_006717081.1 | 28 | 4086 | |||
XM_006717019.1 | XP_006717082.1 | 27 | 4047 | |||
XM_017014552.1 | XP_016870041.1 | 27 | 4017 | |||
GenBank mRNA | AB014574.1 | - | 28 | 4126 | ||
BC026048.1 | - | 3 | 1073 | |||
BC077732.1 | - | 12 | 2409 |