Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015279.1 | NP_056094.1 | 12 | 2660 | ||
XM_011538078.2 | XP_011536380.1 | 13 | 2662 | |||
XM_017019083.1 | XP_016874572.1 | 12 | 2530 | |||
XM_017019084.1 | XP_016874573.1 | 14 | 2321 | |||
XM_017019085.1 | XP_016874574.1 | 14 | 2541 | |||
XM_017019086.1 | XP_016874575.1 | 14 | 2546 | |||
XM_017019087.1 | XP_016874576.1 | 12 | 2412 | |||
GenBank mRNA | AB023201.1 | - | 13 | 2255 |