Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 47 | 6168 | ||
XM_011525642.1 | XP_011523944.1 | 47 | 6168 | |||
XM_011525643.2 | XP_011523945.1 | 47 | 6168 | |||
XM_011525644.2 | XP_011523946.1 | 46 | 6076 | |||
XM_017025684.1 | XP_016881173.1 | 48 | 6346 | |||
GenBank mRNA | AB014550.1 | - | 21 | 2510 | ||
AK025646.1 | - | 8 | 961 | |||
AK126324.1 | - | 31 | 3728 | |||
AL080138.1 | - | 18 | 2148 | |||
CR627458.1 | - | 37 | 4534 |