Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018132.3 | NP_060602.2 | 9 | 817 | ||
XM_005249205.1 | XP_005249262.1 | 9 | 878 | |||
XM_011514723.2 | XP_011513025.2 | 8 | 753 | |||
GenBank mRNA | AA460110.1 | - | 2 | 118 | ||
AK001407.1 | - | 4 | 367 | |||
AK293016.1 | - | 9 | 822 | |||
BC016279.1 | - | 9 | 807 | 3908895 |