Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080398.1 | NP_001073867.1 | 10 | 1436 | ||
XM_005251850.3 | XP_005251907.1 | 8 | 939 | |||
XM_005251851.4 | XP_005251908.1 | 9 | 1318 | |||
XM_005251852.4 | XP_005251909.1 | 9 | 1270 | |||
XM_005251853.3 | XP_005251910.1 | 9 | 1052 | |||
XM_005251854.3 | XP_005251911.1 | 8 | 1221 | |||
XM_011518422.2 | XP_011516724.1 | 8 | 939 | |||
XM_017014557.1 | XP_016870046.1 | 3 | 308 | |||
GenBank mRNA | AK122712.1 | - | 8 | 923 | ||
AK127247.1 | - | 4 | 379 | |||
AK308313.1 | - | 9 | 1043 |