Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080398.1 | NP_001073867.1 | 32 | 3890 | ||
XM_005251850.3 | XP_005251907.1 | 30 | 3393 | |||
XM_005251851.4 | XP_005251908.1 | 31 | 3772 | |||
XM_005251852.4 | XP_005251909.1 | 31 | 3724 | |||
XM_005251853.3 | XP_005251910.1 | 31 | 3506 | |||
XM_005251854.3 | XP_005251911.1 | 30 | 3675 | |||
XM_011518422.2 | XP_011516724.1 | 30 | 3393 | |||
XM_017014557.1 | XP_016870046.1 | 24 | 2712 | |||
GenBank mRNA | AB002366.1 | - | 20 | 2164 | ||
AK024705.1 | - | 7 | 719 | |||
AK025689.1 | - | 15 | 1692 | |||
AK127247.1 | - | 26 | 2831 |