Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080412.2 | NP_001073881.2 | 8 | 1181 | ||
XM_005247256.4 | XP_005247313.1 | 2 | 1701 | |||
XM_005247257.1 | XP_005247314.1 | 3 | 370 | |||
XM_005247258.1 | XP_005247315.1 | 4 | 810 | |||
XM_005247261.3 | XP_005247318.1 | 3 | 439 | |||
XM_011512607.1 | XP_011510909.1 | 4 | 457 | |||
XM_011512608.1 | XP_011510910.1 | 4 | 521 | |||
XM_011512611.1 | XP_011510913.1 | 4 | 462 | |||
XM_017006050.1 | XP_016861539.1 | 5 | 766 | |||
XM_017006051.1 | XP_016861540.1 | 5 | 961 | |||
XM_017006052.1 | XP_016861541.1 | 4 | 884 | |||
XM_017006053.1 | XP_016861542.1 | 5 | 580 | |||
XM_017006054.1 | XP_016861543.1 | 5 | 681 | |||
XM_017006055.1 | XP_016861544.1 | 5 | 585 | |||
XM_017006056.1 | XP_016861545.1 | 5 | 709 | |||
XM_017006057.1 | XP_016861546.1 | 3 | 961 | |||
XM_017006058.1 | XP_016861547.1 | 5 | 604 | |||
XM_017006059.1 | XP_016861548.1 | 4 | 582 | |||
XM_017006060.1 | XP_016861549.1 | 5 | 619 | |||
XM_017006061.1 | XP_016861550.1 | 5 | 560 | |||
GenBank mRNA | AK092355.1 | - | 4 | 478 | ||
AK302568.1 | - | 2 | 249 | |||
AK307452.1 | - | 3 | 545 | |||
BC015444.1 | - | 8 | 1181 |