Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039397.2 | NP_001034486.2 | 10 | 1829 | ||
XM_011523781.2 | XP_011522083.1 | 11 | 1265 | |||
XM_011523782.2 | XP_011522084.1 | 11 | 1145 | |||
XM_017024416.1 | XP_016879905.1 | 10 | 1177 | |||
XM_017024417.1 | XP_016879906.1 | 11 | 1224 | |||
XM_017024418.1 | XP_016879907.1 | 11 | 1322 | |||
XM_017024419.1 | XP_016879908.1 | 10 | 1148 | |||
XM_017024420.1 | XP_016879909.1 | 11 | 2616 | |||
XM_017024421.1 | XP_016879910.1 | 11 | 1195 | |||
XM_017024422.1 | XP_016879911.1 | 10 | 1022 | |||
XM_017024423.1 | XP_016879912.1 | 6 | 854 | |||
GenBank mRNA | AK097563.1 | - | 9 | 2117 | ||
AL137747.1 | - | 1 | 656 |