Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMS1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC131568.1 | - | 1 | 45 |
Gene Symbol: BMS1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008503.1 | - | 1 | 782 |