Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009984.2 | NP_001009984.1 | 34 | 3220 | ||
XM_005260684.3 | XP_005260741.1 | 34 | 3214 | |||
XM_005260687.4 | XP_005260744.1 | 15 | 2056 | |||
XM_011529208.2 | XP_011527510.1 | 15 | 1557 | |||
XM_017027783.1 | XP_016883272.1 | 31 | 2841 | |||
GenBank mRNA | AK226100.1 | - | 29 | 2644 | ||
AK303375.1 | - | 24 | 2500 | |||
AL110249.1 | - | 25 | 2185 | |||
BC064352.1 | - | 8 | 662 | |||
BC080528.1 | - | 9 | 869 | |||
BC122529.1 | - | 25 | 2438 | 40012183 |