Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098816.2 | NP_001092286.2 | 18 | 2892 | ||
XM_017017525.1 | XP_016873014.1 | 17 | 3163 | |||
XM_017017526.1 | XP_016873015.1 | 15 | 4068 | |||
GenBank mRNA | AB037723.3 | - | 3 | 170 |