Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015562.1 | NP_056377.1 | 9 | 969 | ||
XM_011512670.2 | XP_011510972.1 | 9 | 917 | |||
XM_011512671.2 | XP_011510973.1 | 8 | 774 | |||
GenBank mRNA | AB018337.1 | - | 9 | 897 | ||
AK026451.1 | - | 9 | 922 | |||
AK303630.1 | - | 8 | 774 | |||
AK307899.1 | - | 5 | 530 | |||
BC028986.1 | - | 8 | 801 | |||
BC047420.1 | - | 9 | 919 |