Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015562.1 | NP_056377.1 | 11 | 4008 | ||
XM_011512670.2 | XP_011510972.1 | 11 | 3956 | |||
XM_011512671.2 | XP_011510973.1 | 10 | 3813 | |||
GenBank mRNA | AB018337.1 | - | 11 | 3934 | ||
AK026451.1 | - | 12 | 1874 |