Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 8 | 916 | ||
NR_046462.1 | 7 | 879 | ||||
XM_005248480.2 | XP_005248537.1 | 8 | 1051 | |||
XM_011543306.2 | XP_011541608.1 | 8 | 1138 | |||
XM_011543307.2 | XP_011541609.1 | 8 | 1567 | |||
XM_017009332.1 | XP_016864821.1 | 6 | 1814 | |||
XM_017009333.1 | XP_016864822.1 | 6 | 788 | |||
GenBank mRNA | AB020695.1 | - | 8 | 868 | ||
BX640902.1 | - | 7 | 697 | |||
BX647536.1 | - | 7 | 881 | |||
CR936791.1 | - | 8 | 918 |