Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 5 | 682 | ||
XM_011515260.1 | XP_011513562.1 | 5 | 682 | |||
XM_011515262.2 | XP_011513564.1 | 5 | 682 | |||
XM_017011959.1 | XP_016867448.1 | 5 | 682 | |||
XM_017011960.1 | XP_016867449.1 | 5 | 682 | |||
GenBank mRNA | AI935093.1 | - | 3 | 403 | ||
BM553299.1 | - | 2 | 111 |