Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADSYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018161.4 | NP_060631.2 | 21 | 2384 | ||
GenBank mRNA | AK001493.1 | - | 21 | 2325 | ||
AK022436.1 | - | 21 | 2354 | |||
AK094022.1 | - | 12 | 2366 | |||
AK097946.1 | - | 13 | 2263 | |||
BC003638.2 | - | 21 | 2347 | 2966503 | ||
BC003666.2 | - | 21 | 2347 | 2966503 |