Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039960.2 | NP_001035049.1 | 25 | 4912 | ||
NM_001258401.2 | NP_001245330.1 | 25 | 4858 | |||
XM_011539010.1 | XP_011537312.1 | 26 | 4967 | |||
XM_011539011.1 | XP_011537313.1 | 26 | 4912 | |||
XM_011539014.2 | XP_011537316.1 | 16 | 3565 | |||
XM_017020240.1 | XP_016875729.1 | 26 | 4994 | |||
XM_017020242.1 | XP_016875731.1 | 26 | 4981 | |||
GenBank mRNA | AK093005.1 | - | 1 | 1573 | ||
AK093357.1 | - | 4 | 2204 | |||
EU637925.1 | - | 1 | 719 |