Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099650.1 | NP_001093120.1 | 7 | 3996 | ||
NM_173601.1 | NP_775872.1 | 8 | 4089 | |||
XM_017019211.1 | XP_016874700.1 | 8 | 3828 | |||
GenBank mRNA | AK021504.1 | - | 1 | 823 | ||
AK021752.1 | - | 1 | 846 | |||
AL832209.1 | - | 4 | 3226 | |||
BC015597.2 | - | 4 | 3151 |