Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00928 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027074.1 | 3 | 2287 | |||
NR_027075.1 | 4 | 1405 | ||||
NR_027076.1 | 3 | 1118 | ||||
NR_027077.1 | 3 | 1289 | ||||
GenBank mRNA | BC039350.1 | - | 3 | 2288 |