Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042371.2 | NP_001035830.1 | 2 | 762 | ||
GenBank mRNA | BC035985.1 | - | 2 | 726 |