Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018176.3 | NP_060646.2 | 4 | 459 | ||
XM_011513850.2 | XP_011512152.1 | 4 | 361 | |||
XM_017008356.1 | XP_016863845.1 | 4 | 361 | |||
GenBank mRNA | AB067503.1 | - | 4 | 352 | ||
AF467955.1 | - | 4 | 360 | |||
AJ487516.1 | - | 4 | 360 | |||
AJ487958.1 | - | 4 | 382 | |||
AK126093.1 | - | 2 | 73 | |||
AL833647.1 | - | 3 | 150 | |||
BC101759.1 | - | 4 | 459 | 8069265 | ||
BC101761.1 | - | 4 | 459 | 8069267 |