Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136501.2 | NP_001129973.1 | 4 | 964 | ||
NR_134326.1 | 3 | 903 | ||||
XM_017026664.1 | XP_016882153.1 | 3 | 810 | |||
GenBank mRNA | AK027865.1 | - | 1 | 250 | ||
AK098117.1 | - | 4 | 932 | |||
AK299732.1 | - | 4 | 964 | |||
AL832297.1 | - | 4 | 950 | |||
BC125185.1 | - | 3 | 792 | |||
BC125186.1 | - | 4 | 850 | 40122307 |