Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM150B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085488.2 | NP_001078957.1 | 4, 5 | 293 | ||
NM_001282011.1 | NP_001268940.1 | 4, 5 | 342 | |||
XM_005258812.3 | XP_005258869.1 | 6, 7 | 622 | |||
XM_011526850.2 | XP_011525152.1 | 5, 6 | 501 | |||
XM_011526851.2 | XP_011525153.1 | 5, 6 | 499 | |||
XM_011526852.2 | XP_011525154.1 | 5, 6 | 499 | |||
XM_011526853.2 | XP_011525155.1 | 4, 5 | 413 | |||
XM_011526854.2 | XP_011525156.1 | 3, 4 | 578 | |||
XM_011526855.2 | XP_011525157.1 | 5, 6 | 451 | |||
XM_011526856.2 | XP_011525158.1 | 3, 4 | 530 | |||
XM_011526857.2 | XP_011525159.1 | 2, 3 | 193 | |||
XM_017026667.1 | XP_016882156.1 | 5, 6 | 513 | |||
GenBank mRNA | AU311823.1 | - | 4, 5 | 293 | ||
BC171904.1 | - | 4, 5 | 303 | 9054558 |