Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010881.1 | NP_001010881.1 | 3 | 1156 | ||
XM_011541269.2 | XP_011539571.2 | 4 | 1403 | |||
XM_011541271.2 | XP_011539573.2 | 4 | 1252 | |||
XM_011541272.2 | XP_011539574.1 | 4 | 1252 | |||
XM_011541274.2 | XP_011539576.2 | 3 | 1270 | |||
XM_011541275.2 | XP_011539577.2 | 3 | 1067 | |||
XM_011541276.2 | XP_011539578.1 | 4 | 1252 | |||
XM_011541277.2 | XP_011539579.1 | 4 | 1252 | |||
XM_011541278.2 | XP_011539580.1 | 4 | 1252 | |||
XM_017001062.1 | XP_016856551.1 | 5 | 1390 | |||
XM_017001063.1 | XP_016856552.1 | 4 | 1243 | |||
XM_017001064.1 | XP_016856553.1 | 4 | 1252 | |||
GenBank mRNA | AL833920.1 | - | 1 | 18 |