Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018201.4 | NP_060671.3 | 9 | 1025 | ||
XM_005252060.2 | XP_005252117.1 | 8 | 928 | |||
GenBank mRNA | AB209565.1 | - | 6 | 744 | ||
AB449899.1 | - | 9 | 878 | |||
AK027457.1 | - | 1 | 69 | |||
AK123828.1 | - | 5 | 639 | |||
AK289777.1 | - | 9 | 937 | |||
AK295183.1 | - | 6 | 728 | |||
AK307556.1 | - | 5 | 627 | |||
BC014898.1 | - | 9 | 900 | |||
BC132817.1 | - | 9 | 898 | 40146839 | ||
BC136509.1 | - | 9 | 898 | 9020499 | ||
BC150311.1 | - | 9 | 934 | 8860447 |