Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271863.1 | NP_001258792.1 | 9 | 2334 | ||
NM_001271864.1 | NP_001258793.1 | 7 | 1794 | |||
NM_001271865.1 | NP_001258794.1 | 8 | 2009 | |||
NM_018203.2 | NP_060673.1 | 6 | 2122 | |||
GenBank mRNA | AK001610.1 | - | 6 | 2033 | ||
AK096590.1 | - | 9 | 2283 | |||
AK299227.1 | - | 5 | 1345 | |||
BC036076.1 | - | 6 | 1703 | 5302460 |