Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018216.2 | NP_060686.2 | 14, 15 | 1821 | ||
XM_017001633.1 | XP_016857122.1 | 14, 15 | 2234 | |||
GenBank mRNA | AK001644.1 | - | 14, 15 | 1789 | ||
AK223546.1 | - | 14, 15 | 1790 | |||
AK295301.1 | - | 14, 15 | 1821 | |||
AK303127.1 | - | 14, 15 | 1672 | |||
AL442072.1 | - | 14, 15 | 1766 | |||
BC043496.1 | - | 14, 15 | 1784 | 5244472 |