Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145025.1 | NP_001138497.1 | 10 | 1532 | ||
NM_018217.2 | NP_060687.2 | 11 | 1643 | |||
NR_026728.1 | 10 | 1859 | ||||
GenBank mRNA | AF086313.1 | - | 1 | 274 | ||
AK001645.1 | - | 11 | 1625 | |||
AK023931.1 | - | 11 | 1613 | |||
AK096016.1 | - | 9 | 1553 | |||
AK293583.1 | - | 10 | 1514 | |||
AK298794.1 | - | 10 | 1601 | |||
AK300212.1 | - | 10 | 1487 | |||
AK303816.1 | - | 9 | 1285 | |||
AY007088.1 | - | 8 | 1211 | |||
AY358580.1 | - | 10 | 1493 | |||
BC001371.2 | - | 11 | 1610 | 3051376 | ||
BC016184.1 | - | 11 | 1584 | 4860424 | ||
BU188881.1 | - | 1 | 292 | |||
CR457266.1 | - | 11 | 1543 |