Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171795.1 | NP_001165266.1 | 2 | 199 | ||
NM_001171796.1 | NP_001165267.1 | 2 | 199 | |||
NM_001171797.1 | NP_001165268.1 | 2 | 199 | |||
NM_001191035.1 | NP_001177964.1 | 2 | 291 | |||
NM_001191036.1 | NP_001177965.1 | 2 | 146 | |||
XM_017013347.1 | XP_016868836.1 | 2 | 205 | |||
XM_017013349.1 | XP_016868838.1 | 2 | 404 | |||
GenBank mRNA | AK096398.1 | - | 3 | 784 | ||
BC031029.1 | - | 2 | 153 | |||
BC104254.1 | - | 3 | 632 | |||
BC104255.1 | - | 3 | 629 | |||
BC110326.1 | - | 2 | 166 | |||
BC113007.1 | - | 3 | 632 | |||
BC113008.1 | - | 3 | 632 | |||
BX474727.1 | - | 2 | 201 | |||
CB989812.1 | - | 2 | 318 | |||
CR749421.1 | - | 2 | 126 | |||
DA093687.1 | - | 2 | 199 | |||
DA115453.1 | - | 2 | 146 | |||
DQ351291.1 | - | 2 | 171 |