Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161344.1 | NP_001154816.1 | 18 | 2460 | ||
NM_001161345.1 | NP_001154817.1 | 18 | 2457 | |||
NM_001161346.1 | NP_001154818.1 | 18 | 2424 | |||
NM_001161347.1 | NP_001154819.1 | 16 | 2184 | |||
NM_018223.2 | NP_060693.2 | 18 | 2337 | |||
GenBank mRNA | AF170724.1 | - | 18 | 2468 | ||
AK001658.1 | - | 18 | 2317 | |||
AK027687.1 | - | 18 | 2420 | |||
AK054917.1 | - | 4 | 1365 | |||
AK090948.1 | - | 5 | 1435 | |||
AK097671.1 | - | 16 | 2185 | |||
AL137561.1 | - | 1304 | ||||
BC012072.1 | - | 17 | 2350 | 4650348 |