Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308384.1 | NP_001295313.1 | 8 | 2534 | ||
NM_018231.2 | NP_060701.1 | 12 | 2469 | |||
XM_017023393.1 | XP_016878882.1 | 11 | 2950 | |||
XM_017023394.1 | XP_016878883.1 | 9 | 2042 | |||
XM_017023395.1 | XP_016878884.1 | 11 | 2245 | |||
XM_017023396.1 | XP_016878885.1 | 10 | 2097 | |||
GenBank mRNA | AK001677.1 | - | 12 | 2462 | ||
BC001961.1 | - | 11 | 2418 | 3510557 |