Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012241.1 | NP_001012241.1 | 3, 4 | 541 | ||
XM_005257298.4 | XP_005257355.1 | 2, 3 | 2193 | |||
XM_005257299.4 | XP_005257356.3 | 2, 3 | 2193 | |||
GenBank mRNA | BC118997.1 | - | 3, 4 | 464 | 40084759 | |
BC122543.1 | - | 3, 4 | 464 | 40084757 | ||
CR749360.1 | - | 3, 4 | 492 |