Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168499.1 | NP_001161971.1 | 2 | 509 | ||
NM_018235.2 | NP_060705.2 | 3 | 382 | |||
XM_005266728.2 | XP_005266785.1 | 3 | 318 | |||
XM_006722503.2 | XP_006722566.1 | 3 | 179 | |||
XM_011526071.2 | XP_011524373.1 | 3 | 247 | |||
XM_011526072.2 | XP_011524374.1 | 3 | 1202 | |||
XM_011526073.2 | XP_011524375.1 | 2 | 215 | |||
GenBank mRNA | AF258592.1 | - | 2 | 237 | ||
AJ347717.1 | - | 2 | 146 | |||
AK001692.1 | - | 3 | 285 | |||
AK097155.1 | - | 3 | 397 | |||
AK300904.1 | - | 3 | 285 | |||
BC001375.2 | - | 3 | 255 | 3051369 | ||
BC003176.1 | - | 2 | 179 | 2957870 | ||
DC313148.1 | - | 3 | 382 | |||
DC392026.1 | - | 2 | 509 | |||
EU794600.1 | - | 2 | 179 |