Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033887.1 | 12 | 2200 | |||
GenBank mRNA | BC040588.1 | - | 12 | 2188 | ||
BC041010.2 | - | 1 | 701 | |||
BC131526.1 | - | 4 | 1186 |