Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143958.1 | NP_001137430.1 | 1, 2 | 523 | ||
NM_018247.3 | NP_060717.1 | 2, 3 | 631 | |||
GenBank mRNA | AK001718.1 | - | 2, 3 | 500 | ||
AK292823.1 | - | 1, 2 | 388 | |||
AK293225.1 | - | 3, 4 | 456 | |||
AK295567.1 | - | 3, 4 | 452 | |||
AL832490.1 | - | 2, 3 | 554 | |||
AL832815.1 | - | 2, 3 | 493 | |||
BC009006.1 | - | 1, 2 | 374 | 4154200 |