Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB095932.1 | - | 10 | 4433 | ||
BC118982.1 | - | 10 | 2246 | 40026312 |