Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHHADH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038990.1 | 6 | 1054 | |||
GenBank mRNA | BC047598.1 | - | 6 | 1054 |