Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198862.1 | NP_001185791.1 | 3 | 431 | ||
NM_018252.2 | NP_060722.2 | 2 | 272 | |||
XM_005273177.3 | XP_005273234.1 | 2 | 732 | |||
XM_011509717.2 | XP_011508019.1 | 2 | 531 | |||
XM_011509718.2 | XP_011508020.1 | 2 | 561 | |||
XM_017001634.1 | XP_016857123.1 | 2 | 556 | |||
XM_017001635.1 | XP_016857124.1 | 2 | 855 | |||
XM_017001637.1 | XP_016857126.1 | 2 | 855 | |||
GenBank mRNA | AK001736.1 | - | 2 | 242 | ||
AK024066.1 | - | 2 | 250 | |||
AK295224.1 | - | 2 | 262 | |||
AK297200.1 | - | 3 | 431 | |||
BC006320.2 | - | 2 | 231 | 4137890 | ||
CR457268.1 | - | 2 | 104 | |||
DC411913.1 | - | 2 | 272 |