Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024924.1 | NP_001020095.1 | 3 | 469 | ||
NM_018261.3 | NP_060731.2 | 3 | 313 | |||
NM_178237.2 | NP_839955.1 | 3 | 313 | |||
XM_005265747.2 | XP_005265804.1 | 3 | 503 | |||
XM_005265748.1 | XP_005265805.1 | 3 | 315 | |||
XM_005265750.2 | XP_005265807.1 | 3 | 390 | |||
XM_017008405.1 | XP_016863894.1 | 3 | 503 | |||
XM_017008406.1 | XP_016863895.1 | 3 | 315 | |||
XM_017008407.1 | XP_016863896.1 | 3 | 503 | |||
GenBank mRNA | AK001755.1 | - | 3 | 255 | ||
AK027413.1 | - | 3 | 230 | |||
AL833208.1 | - | 2 | 101 | |||
BC094751.1 | - | 3 | 469 | 4828034 | ||
BI760313.1 | - | 3 | 302 | |||
BM468054.1 | - | 3 | 432 |