Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018263.4 | NP_060733.4 | 13 | 3104 | ||
XM_006712039.3 | XP_006712102.1 | 9 | 5225 | |||
XM_006712040.2 | XP_006712103.1 | 9 | 2511 | |||
XM_011532950.2 | XP_011531252.1 | 12 | 3143 | |||
XM_011532951.2 | XP_011531253.1 | 11 | 2831 | |||
XM_017004429.1 | XP_016859918.1 | 13 | 2934 | |||
XM_017004430.1 | XP_016859919.1 | 10 | 5291 | |||
GenBank mRNA | AB051472.2 | - | 13 | 3104 | ||
AB084279.1 | - | 13 | 2967 | |||
AK001760.1 | - | 1 | 217 | |||
BX647527.1 | - | 14 | 3177 | |||
BX648218.1 | - | 13 | 3129 | |||
CR933613.1 | - | 13 | 3148 |