Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277269.1 | NP_001264198.1 | 25 | 3140 | ||
NM_001292063.1 | NP_001278992.1 | 26 | 3104 | |||
GenBank mRNA | AK128214.1 | - | 3 | 469 | ||
DB096404.1 | - | 3 | 470 |