Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277269.1 | NP_001264198.1 | 37 | 6385 | ||
NM_001292063.1 | NP_001278992.1 | 38 | 6349 | |||
GenBank mRNA | AK128214.1 | - | 14 | 3687 |