Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080456.2 | NP_001073925.2 | 4 | 1318 | ||
XM_005258864.3 | XP_005258921.2 | 5 | 1477 | |||
XM_006723189.3 | XP_006723252.1 | 5 | 1498 |