Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512754.1 | XP_011511056.1 | 5, 6 | 964 | ||
XM_011512758.1 | XP_011511060.1 | 4, 5 | 385 |