Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31D5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026851.1 | 4 | 864 | |||
GenBank mRNA | AK125938.1 | - | 4 | 748 | ||
AK128229.1 | - | 4 | 1361 | |||
BC104780.1 | - | 4 | 784 | |||
BC104782.1 | - | 4 | 784 |