Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31D5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026851.1 | 1 | 121 | |||
GenBank mRNA | AK128229.1 | - | 1 | 121 | ||
BC104780.1 | - | 1 | 41 | |||
BC104782.1 | - | 1 | 41 | |||
DB098739.1 | - | 1 | 121 |