Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128223.2 | NP_001121695.1 | 5 | 526 | ||
NM_001290208.1 | NP_001277137.1 | 5 | 507 | |||
NM_001290209.1 | NP_001277138.1 | 5 | 536 | |||
NM_001324027.1 | NP_001310956.1 | 5 | 526 | |||
XM_005264711.3 | XP_005264768.1 | 6 | 604 | |||
XM_006712912.3 | XP_006712975.1 | 5 | 572 | |||
XM_011533244.2 | XP_011531546.1 | 6 | 454 | |||
XM_011533245.2 | XP_011531547.1 | 6 | 590 | |||
XM_011533246.2 | XP_011531548.1 | 6 | 455 | |||
XM_011533248.2 | XP_011531550.1 | 6 | 455 | |||
XM_011533249.2 | XP_011531551.1 | 6 | 453 | |||
GenBank mRNA | DB079386.1 | - | 5 | 536 | ||
DR006008.1 | - | 5 | 582 | |||
HY057162.1 | - | 5 | 472 |