Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC-PINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015431.2 | 5 | 2927 | |||
NR_024153.2 | 5 | 1063 | ||||
NR_109850.1 | 6 | 3051 | ||||
NR_109851.1 | 6 | 3056 | ||||
NR_109852.1 | 6 | 3029 | ||||
NR_109853.1 | 6 | 2097 | ||||
NR_109854.1 | 6 | 2053 | ||||
GenBank mRNA | AK125651.1 | - | 6 | 3030 | ||
AL713664.1 | - | 4 | 1744 | |||
BC039420.1 | - | 5 | 1063 | |||
BC047367.1 | - | 4 | 2073 |