Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GS1-124K5.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111972.1 | 5 | 1574 | |||
NR_111974.1 | 3 | 1299 | ||||
NR_111975.1 | 4 | 1396 | ||||
NR_111976.1 | 4 | 1477 | ||||
NR_111977.1 | 6 | 1643 | ||||
NR_111978.1 | 5 | 1571 | ||||
NR_111979.1 | 4 | 1474 | ||||
GenBank mRNA | AK054923.1 | - | 4 | 2064 | ||
AK126730.1 | - | 4 | 3002 | |||
BC103766.1 | - | 5 | 1543 |